Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Brd4 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A308055-100 A308055-50
77228-420EA 494.4 USD InStock
77228-420 77228-422
Anti-Brd4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Brd4 for WB with samples derived from Human and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: Brd4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty