Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PI3 Kinase p110 beta Rabbit Monoclonal Antibody [clone: ARC0703]

Supplier: ANTIBODIES.COM LLC
A309440-100
77215-260EA 494.4 USD InStock
77215-260 77205-186
Anti-PI3 Kinase p110 beta Rabbit Monoclonal Antibody [clone: ARC0703]
Antibodies
Rabbit monoclonal [ARC0703] antibody to PI3 Kinase p110 beta for WB and IHC with samples derived from Human and Mouse.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:1,000, IHC: 1:50 to 1:200

Type: Primary
Antigen: PI3 Kinase p110 beta
Clonality: Monoclonal
Clone: ARC0703
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty