Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD47 Rabbit Recombinant Antibody [clone: DM212]

Supplier: ANTIBODIES.COM LLC
A318497-100 A318497-500
77186-626EA 703.3 USD InStock
77186-626 77186-628
Anti-CD47 Rabbit Recombinant Antibody [clone: DM212]
Antibodies
Recombinant rabbit monoclonal [DM212] antibody to CD47 for ELISA and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: CD47
Clonality: Recombinant
Clone: DM212
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty