Anti-Spa-1 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A305800-100 A305800-50
77219-514EA 470 USD
77219-514 77219-516
Anti-Spa-1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Spa-1 for WB and IHC with samples derived from Human and Mouse.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:100 to 1:200

Type: Primary
Antigen: Spa-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now