Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-AMPK alpha 1 (phospho Thr183) + AMPK alpha 2 (phospho Thr172) (phospho Thr183/phospho Thr172) Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A306718-100
77223-138EA 351.7 USD InStock
77223-138 77223-140
Anti-AMPK alpha 1 (phospho Thr183) + AMPK alpha 2 (phospho Thr172) (phospho Thr183/phospho Thr172) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AMPK alpha 1 (phospho Thr183) + AMPK alpha 2 (phospho Thr172) for WB with samples derived from Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: AMPK alpha 1 (phospho Thr183) + AMPK alpha 2 (phospho Thr172)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty