Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-beta Tubulin Mouse Monoclonal Antibody [clone: BT7R]

Supplier: ANTIBODIES.COM LLC
A85273-100 A85273-200
77064-774EA 397.5 USD InStock
77064-774 77080-792
Anti-beta Tubulin Mouse Monoclonal Antibody [clone: BT7R]
Antibodies
Mouse monoclonal (BT7R) antibody tobeta Tubulin for Dot, ELISA, IS and WB with samples derived from Chicken, Human, Monkey, Mouse, Rat and Rabbit.

  • Validated Applications: Dot, ELISA, IS, W

Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: BT7R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken;Human;Monkey;Mouse;Rat;Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty