Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-alpha Internexin Mouse Monoclonal Antibody [clone: 2E3]

Supplier: ANTIBODIES.COM LLC
A85448-250 A85448-500
77076-782EA 428.1 USD InStock
77076-782 77081-342
Anti-alpha Internexin Mouse Monoclonal Antibody [clone: 2E3]
Antibodies
Mouse monoclonal (2E3) antibody to alpha Internexin for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse and Bovine.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:10000, ICC/IF: 1:5000, IHC: 1:500

Type: Primary
Antigen: alpha Internexin
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse;Bovine

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty