Burrowing Tubes
Supplier: MAZE ENGINEERS
4915 4916 4917 4918 4919 4920
76670-166EA 253.34 USD
76670-166 76670-168 76670-170 76670-172 76670-174 76670-176
Burrowing Tubes
Animal Supplies Animal Environmental Enrichment
The Burrowing tube studies burrowing behaviour in which rodents spontaneously empty a tube filled with food pellets, gravel, or other substances. This is a sensitive test to various pathologies including prion diseases and inflammatory conditions.
  • Acrylic construction
  • Non reflective surfaces
  • Easy to clean
The Burrowing tube is manufactured from acrylic, with acrylic tubing and a 1 cm fiberboard backing that is sealed with a waterproof adhesive. Each tube includes legging on the proximal side for elevation off of the floor.