Immunotag™ Anti-STAB1 Rabbit Polyclonal Antibody (Alexa Fluor® 405)

Supplier: G-Biosciences
ITN2164100U405 ITN216450U405
77322-226EA 524.42 USD
77322-226 77322-246
Immunotag™ Anti-STAB1 Rabbit Polyclonal Antibody (Alexa Fluor® 405)
Antibodies
Encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 FAS1, 16 EGF-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluronan-binding link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node. The receptor has been shown to endocytose ligands such as low-density lipoprotein, gram-positive and gram-negative bacteria, and advanced glycosylation end products. Supporting its possible role as a scavenger receptor, the protein rapidly cycles between the plasma membrane and early endosomes. [provided by RefSeq, Jul 2008].

Acts as a scavenger receptor for acetylated low density lipoprotein. Binds to both gram-positive and gram-negative bacteria and may play a role in defense against bacterial infection. When inhibited in endothelial tube formation assays, there is a marked decrease in cell-cell interactions, suggesting a role in angiogenesis. Involved in the delivery of newly synthesized CHID1/SI-CLP from the biosynthetic compartment to the endosomal/lysosomal system.

Type: Primary
Antigen: STAB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 405
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR