2-Bromo-4-(4,4-dimethylcyclohexyl)phenol ≥97%

Supplier: AOB CHEM USA
Warning

109971-500MG 109971-1G 109971-5G 109971-25G
77323-464EA 271.92 USD
77323-464 77347-532 77359-574 77383-476
2-Bromo-4-(4,4-dimethylcyclohexyl)phenol ≥97%
2-Bromo-4-(4,4-dimethylcyclohexyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₁₉BrO
MW: 283.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780375
CAS Number: 2901110-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR