Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluorophenyl acetate

Supplier: AOB CHEM USA
Warning

111205-500MG 111205-1G 111205-5G 111205-25G
77323-480EA 105.06 USD InStock
77323-480 77343-104 77359-590 77371-564
2,4-Difluorophenyl acetate
2,4-Difluorophenyl acetate
Precaución: for Research Use Only


Formula: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00116503
CAS Number: 36914-77-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty