4-(tert-Butyl)-2-isopropoxybenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

112841-500MG 112841-1G 112841-5G
77337-686EA 435.69 USD
77337-686 77347-570 77371-598
4-(tert-Butyl)-2-isopropoxybenzaldehyde ≥97%
4-(tert-Butyl)-2-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
MW: 220.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29121488
CAS Number: 1289162-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR