Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,6-difluoronicotinate ≥97%

Supplier: AOB CHEM USA
Warning

18655-500MG 18655-1G 18655-5G 18655-25G
77324-182EA 96.82 USD InStock
77324-182 77342-640 77360-454 77372-378
Ethyl 2,6-difluoronicotinate ≥97%
Ethyl 2,6-difluoronicotinate
Precaución: for Research Use Only


Formula: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD14698230
CAS Number: 939812-72-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty