Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-(4-fluoro-3-methylthiophenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

26464-500MG 26464-1G 26464-5G
77324-800EA 345.05 USD InStock
77324-800 77348-648 77372-888
Ethyl 4-(4-fluoro-3-methylthiophenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate ≥97%
Ethyl 4-(4-fluoro-3-methylthiophenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Precaución: for Research Use Only


Formula: C₁₅H₁₇FN₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD27756659
CAS Number: 1809161-47-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty