Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-(1-methylethyl)-benzoic acid ethyl ester ≥97%

Supplier: AOB CHEM USA
Warning

29514-500MG 29514-1G 29514-5G
77336-328EA 387.28 USD InStock
77336-328 77348-832 77373-102
4-Chloro-3-(1-methylethyl)-benzoic acid ethyl ester ≥97%
4-Chloro-3-(1-methylethyl)-benzoic acid ethyl ester
Precaución: for Research Use Only


Formula: C₁₂H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD31619425
CAS Number: 1008529-62-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty