Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dibromothiophene-2-boronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

32721-500MG 32721-1G 32721-5G 32721-25G
77325-384EA 228.66 USD InStock
77325-384 77349-128 77361-744 77384-848
3,4-Dibromothiophene-2-boronic acid pinacol ester ≥97%
3,4-Dibromothiophene-2-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₀H₁₃BBr₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD30529851
CAS Number: 2121513-10-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty