Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(methoxymethyl)benzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

36478-500MG 36478-1G 36478-5G 36478-25G
77326-124EA 218.36 USD InStock
77326-124 77349-822 77362-438 77385-366
2-Fluoro-5-(methoxymethyl)benzoic acid ≥95%
2-Fluoro-5-(methoxymethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD32127075
CAS Number: 1697993-35-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty