Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3,5-dichloro-2-fluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

37380-500MG 37380-1G 37380-5G 37380-25G
77326-360EA 236.9 USD InStock
77326-360 77350-058 77362-666 77385-516
tert-Butyl 3,5-dichloro-2-fluorobenzoate ≥97%
tert-Butyl 3,5-dichloro-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₁Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD34564045
CAS Number: 2734775-94-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty