Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-chloro-2-ethoxy-6-fluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

47966-500MG 47966-1G 47966-5G
77327-418EA 342.99 USD InStock
77327-418 77350-986 77375-332
Methyl 3-chloro-2-ethoxy-6-fluorobenzoate ≥97%
Methyl 3-chloro-2-ethoxy-6-fluorobenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34756390
CAS Number: 2385879-42-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty