Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,6-difluoro-4-(methylthio)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

48393-500MG 48393-1G 48393-25G 48393-5G
77327-562EA 291.49 USD InStock
77327-562 77351-158 77386-338 77375-464
Methyl 2,6-difluoro-4-(methylthio)benzoate ≥97%
Methyl 2,6-difluoro-4-(methylthio)benzoate
Precaución: for Research Use Only


Formula: C₉H₈F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34564117
CAS Number: 2734772-68-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty