Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-ethoxynaphthalene ≥97%

Supplier: AOB CHEM USA
Warning

57144-500MG 57144-1G 57144-5G 57144-25G
77329-858EA 85.49 USD InStock
77329-858 77342-220 77366-154 77378-056
2-Bromo-1-ethoxynaphthalene ≥97%
2-Bromo-1-ethoxynaphthalene
Precaución: for Research Use Only


Formula: C₁₂H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD34758086
CAS Number: 108238-18-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty