3,4-Difluoro-1,1'-biphenyl ≥97%

Supplier: AOB CHEM USA
Warning

61696-500MG 61696-1G 61696-5G 61696-25G
77331-358EA 131.84 USD
77331-358 77344-374 77367-498 77389-002
3,4-Difluoro-1,1'-biphenyl ≥97%
3,4-Difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈F₂
MW: 190.1887 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701761
CAS Number: 67277-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR