Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-5-methylphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOB CHEM USA
Warning

62315-500MG 62315-1G 62315-5G 62315-25G
77331-600EA 126.69 USD InStock
77331-600 77344-178 77367-730 77379-890
(3-Bromo-5-methylphenyl)(thiomorpholino)methanone ≥97%
(3-Bromo-5-methylphenyl)(thiomorpholino)methanone
Precaución: for Research Use Only


Formula: C₁₂H₁₄BrNOS
Storage Temperature: Ambient
MDL Number: MFCD34760797
CAS Number: 1871083-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty