Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methyl-1-naphthoic acid ≥97%

Supplier: AOB CHEM USA
Warning

65132-500MG 65132-1G 65132-5G
77339-226EA 568.56 USD InStock
77339-226 77356-770 77380-714
4-Fluoro-3-methyl-1-naphthoic acid ≥97%
4-Fluoro-3-methyl-1-naphthoic acid
Precaución: for Research Use Only


Formula: C₁₂H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD34762112
CAS Number: 2807466-11-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty