3-Chloro-6-iodo-2-(methylthio)phenol ≥97%

Supplier: AOB CHEM USA
Warning

74487-500MG 74487-1G 74487-5G
77337-616EA 428.48 USD
77337-616 77358-206 77382-108
3-Chloro-6-iodo-2-(methylthio)phenol ≥97%
3-Chloro-6-iodo-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆ClIOS
MW: 300.544 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765510
CAS Number: 2710589-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR