Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isobutoxy-2,5-dimethylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

76238-500MG 76238-1G 76238-5G 76238-25G
77334-210EA 202.91 USD InStock
77334-210 77358-402 77370-216 77391-020
4-Isobutoxy-2,5-dimethylbenzoic acid ≥97%
4-Isobutoxy-2,5-dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD28580805
CAS Number: 1368030-30-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty