4-Chloro-5-fluoro-2-(methylthio)phenol ≥97%
Supplier: AOB CHEM USA
|
Warning
|
94178-500MG
94178-1G
94178-5G
94178-25G
77335-190EA
252.35
USD
77335-190
77359-280
77371-232
77391-698
4-Chloro-5-fluoro-2-(methylthio)phenol ≥97%
4-Chloro-5-fluoro-2-(methylthio)phenol
Precaución: for Research Use Only
Formula:
C₇H₆ClFOS MW: 192.638 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34773938 CAS Number: 2710589-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...