2-(2,2-Dichloro-1-hydroxyethyl)phenol ≥95%

Supplier: AOB CHEM USA
Warning

54993-5G 54993-25G
77365-666EA 166.86 USD
77365-666 77377-494
2-(2,2-Dichloro-1-hydroxyethyl)phenol ≥95%
2-(2,2-Dichloro-1-hydroxyethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O₂
MW: 207.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757426
CAS Number: 2755723-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR