Tungsten trioxide 99+%

Supplier: APOLLO SCIENTIFIC
Warning

IN3736-100G IN3736-500G
77401-602EA 151.8 USD
77401-602 77401-604
Tungsten trioxide 99+%
Tungsten trioxide
Formula: O₃W
MW: 231.84
Melting Pt: 1473
Density: 7.16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011466
CAS Number: 1314-35-8
EINECS: 215-231-4
Merck Index: 12,09947

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR