Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indole-2-carbonyl chloride

Supplier: APOLLO SCIENTIFIC

OR111006-1G OR111006-5G
77403-614EA 227.2 USD InStock
77403-614 77403-616
1H-Indole-2-carbonyl chloride
1H-Indole-2-carbonyl chloride


Formula: C₉H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD09833544
CAS Number: 58881-45-1
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty