Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[b]thiophene-7-carbonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR12299-1G OR12299-250MG
77404-252EA 209.88 USD InStock
77404-252 77404-254
Benzo[b]thiophene-7-carbonyl chloride 97%
Benzo[b]thiophene-7-carbonyl chlorid


Formula: C₉H₅ClOS
Melting Pt: 84…86 °C
Storage Temperature: Ambient
CAS Number: 120081-47-2
UN: 1759
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty