Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazine-2-carbaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR1854-1G OR1854-250MG OR1854-5G
77406-340EA 136.52 USD InStock
77406-340 77406-342 77406-344
Pyrazine-2-carbaldehyde 97%
Pyrazine-2-carbaldehyde


Formula: C5H4N2O
Boiling Pt: 120…130 °C/30 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD02179278
CAS Number: 5780-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty