Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis,cis-9,12-Octadecadien-1-ol 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR19496-1ML OR19496-5ML
77406-470EA 74.38 USD InStock
77406-470 77406-472
cis,cis-9,12-Octadecadien-1-ol 95%
cis,cis-9,12-Octadecadien-1-ol


Formula: C₁₈H₃₄O
Boiling Pt: 155 °C/0.5 mmHg
Density: 0.86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00056667
CAS Number: 506-43-4
EINECS: 208-038-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty