Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Diaminoanisole

Supplier: APOLLO SCIENTIFIC
Warning

OR21113-1G OR21113-25G OR21113-5G
77406-618EA 21.26 USD InStock
77406-618 77406-620 77406-622
3,4-Diaminoanisole
3,4-Diaminoanisole


Formula: C₇H₁₀N₂O
Melting Pt: 46…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047837
CAS Number: 102-51-2
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty