2-Mercaptonicotinic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR28620-100G OR28620-25G OR28620-500G
77408-120EA 57.06 USD
77408-120 77408-122 77408-124
2-Mercaptonicotinic acid 97%
2-Mercaptonicotinic acid
Formula: C₆H₅NO₂S
MW: 155.18 g/mol
Melting Pt: 263…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00010102
CAS Number: 38521-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR