Dihydrolinalool 95%

Supplier: APOLLO SCIENTIFIC

OR931110-100G OR931110-25G
77411-960EA 204.78 USD
77411-960 77411-962
Dihydrolinalool 95%
Dihydrolinalool
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059140
CAS Number: 18479-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR