Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S)-3-(Trifluoromethyl)piperidine 97%

Supplier: APOLLO SCIENTIFIC

PC520665-1G PC520665-500MG
77416-516EA 478.84 USD InStock
77416-516 77416-520
(3S)-3-(Trifluoromethyl)piperidine 97%
(3S)-3-(Trifluoromethyl)piperidine


Formula: C₆H₁₀F₃N
Storage Temperature: Ambient
MDL Number: MFCD11975307
CAS Number: 749208-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty