Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7294-25G PC7294-5G
77417-774EA 149.76 USD InStock
77417-774 77417-776
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione 98%
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione


Formula: C₈H₁₁F₃O₂
Boiling Pt: 138…141 °C
Density: 1.129 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00040841
CAS Number: 22767-90-4
EINECS: 245-211-0
UN: 1224
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty