Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α'-Bromo-α,α,α-trifluoro-o-xylene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7631-100G PC7631-25G PC7631-5G
77418-044EA 93.74 USD InStock
77418-044 77418-046 77418-048
α'-Bromo-α,α,α-trifluoro-o-xylene 98%
α'-Bromo-α,α,α-trifluoro-o-xylene


Formula: C₈H₆BrF₃
Boiling Pt: 70…72 °C/7 mmHg
Melting Pt: 27…30 °C
Density: 1.571 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00013558
CAS Number: 395-44-8
EINECS: 206-901-7
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty