Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetrafluoro-1-propanol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC6864-100G PC6864-1KG PC6864-250G
77417-568EA 151.8 USD InStock
77417-568 77417-570 77417-572
2,2,3,3-Tetrafluoro-1-propanol 98%
2,2,3,3-Tetrafluoro-1-propanol
  • Surface tension 27.6 dynes/cm 200C
  • Dissociation constant Ka=4.6 x 10-12


Formula: C₃H₄F₄O
MW: 132.06 g/mol
Boiling Pt: 109
Melting Pt: -15
Density: 1.471 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00004676
CAS Number: 76-37-9
EINECS: 200-955-5
UN: 1986
ADR: 3+6.1 III
Merck Index: 12,09346
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty