Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diaminopyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR13639-100G OR13639-500G
77404-694EA 96.78 USD InStock
77404-694 77404-696
2,6-Diaminopyridine
2,6-Diaminopyridine


Formula: C₅H₇N₃
Boiling Pt: 285 °C
Melting Pt: 117…122 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006329
CAS Number: 141-86-6
EINECS: 205-507-2
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty