Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Difluoroiodomethane 10% (w/w) in THF

Supplier: APOLLO SCIENTIFIC
Danger

PC49177-10G PC49177-25G
77415-878EA 241.46 USD InStock
77415-878 77415-880
Difluoroiodomethane 10% (w/w) in THF
Difluoroiodomethane


Formula: CHF₂I
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD03094113
CAS Number: 1493-03-4
UN: 2056
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty