Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluoro-2-(trifluoromethyl)aniline 97%

Supplier: APOLLO SCIENTIFIC

PC302856-1G PC302856-5G
77414-296EA 172.18 USD InStock
77414-296 77414-298
4-Bromo-3-fluoro-2-(trifluoromethyl)aniline 97%
4-Bromo-3-fluoro-2-(trifluoromethyl)aniline


Formula: C₇H₄BrF₄N
Storage Temperature: Ambient
MDL Number: MFCD11519308
CAS Number: 1092460-59-7
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty