tert-Butyl-3-aminobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

OR2417-1G OR2417-25G OR2417-5G
77407-436EA 117.16 USD
77407-436 77407-438 77407-440
tert-Butyl-3-aminobenzoate
tert-Butyl-3-aminobenzoate
Formula: C₁₁H₁₅NO₂
MW: 193.2423 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00729048
CAS Number: 92146-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR