Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-5-nitroaniline

Supplier: APOLLO SCIENTIFIC
Warning

PC2673-1G PC2673-25G PC2673-5G
77413-744EA 27.4 USD InStock
77413-744 77413-746 77413-748
4-Bromo-2-fluoro-5-nitroaniline
4-Bromo-2-fluoro-5-nitroaniline


Formula: C₆H₄BrFN₂O₂
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD08741367
CAS Number: 87547-06-6
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty