Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Hydroxyethyl)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR30086-1G OR30086-250MG
77408-704EA 33.62 USD InStock
77408-704 77408-706
4-(2-Hydroxyethyl)benzonitrile 98%
4-(2-Hydroxyethyl)benzonitrile


Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 116…118 °C/0.5 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052461
CAS Number: 69395-13-7
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty