Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-2-oxo-1,2-dihydroquinoline-4-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR13262-1G OR13262-250MG OR13262-500MG
77404-610EA 316.78 USD InStock
77404-610 77404-612 77404-614
1-Methyl-2-oxo-1,2-dihydroquinoline-4-carbaldehyde
1-Methyl-2-oxo-1,2-dihydroquinoline-4-carbaldehyde


Formula: C₁₁H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD03942175
CAS Number: 15112-98-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty