Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Morpholinonicotinoyl chloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR23271-10G OR23271-1G OR23271-5G
77407-206EA 578.68 USD InStock
77407-206 77407-208 77407-210
6-Morpholinonicotinoyl chloride, Technical Grade
6-Morpholinonicotinoyl chloride


Formula: C₁₀H₁₁ClN₂O₂
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD02681966
CAS Number: 313350-36-6
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty