Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate

Supplier: APOLLO SCIENTIFIC

OR110188-1G OR110188-5G
77403-114EA 143.66 USD InStock
77403-114 77403-116
Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate
Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate


Formula: C₁₁H₁₁ClO₃S
Storage Temperature: Ambient
MDL Number: MFCD11100102
CAS Number: 1215582-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty