Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate

Supplier: APOLLO SCIENTIFIC

OR110207-5G OR110207-1G
77403-134EA 199.7 USD InStock
77403-134 77403-132
Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate
Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate


Formula: C₁₁H₁₀BrNO₂S
Storage Temperature: Ambient
MDL Number: MFCD11505028
CAS Number: 1216708-87-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty